missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl Trifluoropyruvate 98.0+%, TCI America™
$28.21 - $69.68
Chemical Identifiers
CAS | 13089-11-7 |
---|---|
Molecular Formula | C4H3F3O3 |
Molecular Weight (g/mol) | 156.06 |
MDL Number | MFCD00114936 |
InChI Key | XGLLQDIWQRQROJ-UHFFFAOYSA-N |
Synonym | methyl trifluoropyruvate, methyl 3,3,3-trifluoropyruvate, methyltrifluoropyruvate, trifluoropyruvic acid methyl ester, 3,3,3-trifluoropyruvic acid methyl ester, propanoic acid, 3,3,3-trifluoro-2-oxo-, methyl ester, pubchem2012, rarechem al bf 1300, acmc-1bp0j |
PubChem CID | 2734931 |
IUPAC Name | methyl 3,3,3-trifluoro-2-oxopropanoate |
SMILES | COC(=O)C(=O)C(F)(F)F |
Chemical Identifiers
13089-11-7 | |
156.06 | |
XGLLQDIWQRQROJ-UHFFFAOYSA-N | |
2734931 | |
COC(=O)C(=O)C(F)(F)F |
C4H3F3O3 | |
MFCD00114936 | |
methyl trifluoropyruvate, methyl 3,3,3-trifluoropyruvate, methyltrifluoropyruvate, trifluoropyruvic acid methyl ester, 3,3,3-trifluoropyruvic acid methyl ester, propanoic acid, 3,3,3-trifluoro-2-oxo-, methyl ester, pubchem2012, rarechem al bf 1300, acmc-1bp0j | |
methyl 3,3,3-trifluoro-2-oxopropanoate |
Specifications
13089-11-7 | |
86°C | |
MFCD00114936 | |
3272 | |
XGLLQDIWQRQROJ-UHFFFAOYSA-N | |
methyl 3,3,3-trifluoro-2-oxopropanoate | |
2734931 | |
≥98.0% (GC) | |
Methyl Trifluoropyruvate |
Yellow | |
C4H3F3O3 | |
1 g | |
methyl trifluoropyruvate, methyl 3,3,3-trifluoropyruvate, methyltrifluoropyruvate, trifluoropyruvic acid methyl ester, 3,3,3-trifluoropyruvic acid methyl ester, propanoic acid, 3,3,3-trifluoro-2-oxo-, methyl ester, pubchem2012, rarechem al bf 1300, acmc-1bp0j | |
COC(=O)C(=O)C(F)(F)F | |
156.06 | |
156.06 | |
Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator