Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl trifluoropyruvate, 97%

$73.69 - $73.69

Chemical Identifiers

Chemical Name or Material Methyl trifluoropyruvate
CAS 13089-11-7
Molecular Formula C4H3F3O3
Formula Weight 156.06
MDL Number MFCD00114936
UN Number UN3272
Beilstein 1863490
Synonym Trifluoropyruvic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2158906

  SDS
alfa aesar™
B2158906
5g Each for $73.69

AAB2158903

  SDS
alfa aesar™
B2158903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2158914

  SDS
alfa aesar™
B2158914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 156.06
Synonym Trifluoropyruvic acid methyl ester
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Boiling Point 84° to 86°C
Flash Point 26°C (79°F)
Health Hazard 1 Warning
Assay 97%
Molecular Formula C4H3F3O3
MDL Number MFCD00114936
Beilstein 1863490
Chemical Name or Material Methyl trifluoropyruvate
UN Number UN3272
CAS 13089-11-7
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.