missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Tridecafluoroheptanoate, TCI America™

$56.53 - $175.70

Chemical Identifiers

CAS 14312-89-1
Molecular Formula C8H3F13O2
Molecular Weight (g/mol) 378.089
MDL Number MFCD00153212
InChI Key JHROQORAJUWVCD-UHFFFAOYSA-N
Synonym Methyl Perfluoroheptanoate, Tridecafluoroheptanoic Acid Methyl Ester, Perfluoroheptanoic Acid Methyl Ester
PubChem CID 526471
IUPAC Name methyl 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptanoate
SMILES COC(=O)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M191425G View Documents TCI America
M191425G
25g Each for $175.70
 
M19145G View Documents TCI America
M19145G
5g Each for $56.53
 
Specifications

Chemical Identifiers

14312-89-1
378.089
JHROQORAJUWVCD-UHFFFAOYSA-N
526471
COC(=O)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
C8H3F13O2
MFCD00153212
Methyl Perfluoroheptanoate, Tridecafluoroheptanoic Acid Methyl Ester, Perfluoroheptanoic Acid Methyl Ester
methyl 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptanoate

Specifications

141°C
25g
C8H3F13O2
1993
JHROQORAJUWVCD-UHFFFAOYSA-N
methyl 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptanoate
526471
Clear Liquid at 20°C
Methyl Tridecafluoroheptanoate
Colorless
14312-89-1
MFCD00153212
Methyl Perfluoroheptanoate, Tridecafluoroheptanoic Acid Methyl Ester, Perfluoroheptanoic Acid Methyl Ester
COC(=O)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
378.089
378.09
≥96.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No