missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Tricosafluorododecanoate 95.0+%, TCI America™

$79.06 - $267.83

Chemical Identifiers

CAS 56554-52-0
Molecular Formula C13H3F23O2
Molecular Weight (g/mol) 628.128
MDL Number MFCD00236616
InChI Key ABNZIVVYHQYSLI-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoic acid methyl ester, acmc-1b030, dodecanoic acid, tricosafluoro-, methyl ester, methyl perfluorododecanoate, methyl tricosafluorododecanoate, methyl tricosafluorolaurate, methyl tricosafluorolaurate; perfluorododecanoic acid methyl ester, perfluorododecanoic acid methyl ester, tricosafluorododecanoic acid methyl ester
PubChem CID 547885
IUPAC Name methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoate
SMILES COC(=O)C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M19175G View Documents Tci America
M19175G
5g Each for $79.06
 
M191725G View Documents Tci America
M191725G
25g Each for $267.83
 
Specifications

Chemical Identifiers

56554-52-0
628.128
ABNZIVVYHQYSLI-UHFFFAOYSA-N
547885
COC(=O)C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
C13H3F23O2
MFCD00236616
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoic acid methyl ester, acmc-1b030, dodecanoic acid, tricosafluoro-, methyl ester, methyl perfluorododecanoate, methyl tricosafluorododecanoate, methyl tricosafluorolaurate, methyl tricosafluorolaurate; perfluorododecanoic acid methyl ester, perfluorododecanoic acid methyl ester, tricosafluorododecanoic acid methyl ester
methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoate

Specifications

125°C
58°C
56554-52-0
MFCD00236616
ABNZIVVYHQYSLI-UHFFFAOYSA-N
methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoate
547885
Crystal-Powder at 20°C
Methyl Tricosafluorododecanoate
White-Yellow
5g
C13H3F23O2
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoic acid methyl ester, acmc-1b030, dodecanoic acid, tricosafluoro-, methyl ester, methyl perfluorododecanoate, methyl tricosafluorododecanoate, methyl tricosafluorolaurate, methyl tricosafluorolaurate; perfluorododecanoic acid methyl ester, perfluorododecanoic acid methyl ester, tricosafluorododecanoic acid methyl ester
COC(=O)C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
628.128
628.13
≥95.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No