missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl trans,trans-9,12-Octadecadienoate, TCI America™

$67.22 - $233.75

Chemical Identifiers

CAS 2566-97-4
Molecular Formula C19H34O2
Molecular Weight (g/mol) 294.479
MDL Number MFCD00069997
InChI Key WTTJVINHCBCLGX-ZDVGBALWSA-N
Synonym methyl linolelaidate, methyl 9e,12e-octadeca-9,12-dienoate, methyl trans,trans-9,12-octadecadienoate, unii-jrb7acq5nw, methyl linoleate hydroperoxide, linolelaidic acid, methyl ester, methyl octadeca-9,12-dienoate, jrb7acq5nw, 9,12-octadecadienoic acid, methyl ester, e,e, linolelaidic acid methyl ester
PubChem CID 5362793
IUPAC Name methyl (9E,12E)-octadeca-9,12-dienoate
SMILES CCCCCC=CCC=CCCCCCCCC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M2307500MG View Documents TCI America
M2307500MG
500mg Each for $233.75
 
M2307100MG View Documents TCI America
M2307100MG
100mg Each for $67.22
 
Specifications

Chemical Identifiers

2566-97-4
294.479
WTTJVINHCBCLGX-ZDVGBALWSA-N
5362793
CCCCCC=CCC=CCCCCCCCC(=O)OC
C19H34O2
MFCD00069997
methyl linolelaidate, methyl 9e,12e-octadeca-9,12-dienoate, methyl trans,trans-9,12-octadecadienoate, unii-jrb7acq5nw, methyl linoleate hydroperoxide, linolelaidic acid, methyl ester, methyl octadeca-9,12-dienoate, jrb7acq5nw, 9,12-octadecadienoic acid, methyl ester, e,e, linolelaidic acid methyl ester
methyl (9E,12E)-octadeca-9,12-dienoate

Specifications

152°C
500mg
C19H34O2
methyl linolelaidate, methyl 9e,12e-octadeca-9,12-dienoate, methyl trans,trans-9,12-octadecadienoate, unii-jrb7acq5nw, methyl linoleate hydroperoxide, linolelaidic acid, methyl ester, methyl octadeca-9,12-dienoate, jrb7acq5nw, 9,12-octadecadienoic acid, methyl ester, e,e, linolelaidic acid methyl ester
CCCCCC=CCC=CCCCCCCCC(=O)OC
294.479
294.48
≥98.0% (GC)
Colorless
2566-97-4
MFCD00069997
WTTJVINHCBCLGX-ZDVGBALWSA-N
methyl (9E,12E)-octadeca-9,12-dienoate
5362793
Clear Liquid at 20°C
Methyl trans,trans-9,12-Octadecadienoate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0798&(2)-1085

TSCA : No

Recommended Storage : Freezer