missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl trans-4-Hydroxycyclohexanecarboxylate, TCI America™

$139.03 - $440.02

Chemical Identifiers

CAS 6125-57-1
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
InChI Key HYDYVXROZHFTGB-UHFFFAOYSA-N
Synonym trans-4-Hydroxycyclohexanecarboxylic Acid Methyl Ester
PubChem CID 87117
IUPAC Name methyl 4-hydroxycyclohexane-1-carboxylate
SMILES COC(=O)C1CCC(CC1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M19911G View Documents TCI America
M19911G
1g Each for $139.03
 
M19915G View Documents TCI America
M19915G
5g Each for $440.02
 
Specifications

Chemical Identifiers

6125-57-1
158.197
trans-4-Hydroxycyclohexanecarboxylic Acid Methyl Ester
methyl 4-hydroxycyclohexane-1-carboxylate
C8H14O3
HYDYVXROZHFTGB-UHFFFAOYSA-N
87117
COC(=O)C1CCC(CC1)O

Specifications

125°C
1g
C8H14O3
HYDYVXROZHFTGB-UHFFFAOYSA-N
methyl 4-hydroxycyclohexane-1-carboxylate
87117
Clear Liquid at 20°C
Methyl trans-4-Hydroxycyclohexanecarboxylate
Yellow
6125-57-1
trans-4-Hydroxycyclohexanecarboxylic Acid Methyl Ester
COC(=O)C1CCC(CC1)O
158.197
158.20
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No