missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl trans-4-(Aminomethyl)cyclohexanecarboxylate Hydrochloride, TCI America™

$113.06 - $368.20

Chemical Identifiers

CAS 29275-88-5
Molecular Formula C9H18ClNO2
Molecular Weight (g/mol) 207.698
MDL Number MFCD00797465
InChI Key PZPSRTWFJCGJLP-UHFFFAOYSA-N
Synonym trans-4-(Aminomethyl)cyclohexanecarboxylic Acid Methyl Ester Hydrochloride
PubChem CID 12721445
IUPAC Name methyl 4-(aminomethyl)cyclohexane-1-carboxylate;hydrochloride
SMILES COC(=O)C1CCC(CC1)CN.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M19885G View Documents TCI America
M19885G
5g Each for $113.06
 
M198825G View Documents TCI America
M198825G
25g Each for $368.20
 
Specifications

Chemical Identifiers

29275-88-5
207.698
PZPSRTWFJCGJLP-UHFFFAOYSA-N
12721445
COC(=O)C1CCC(CC1)CN.Cl
C9H18ClNO2
MFCD00797465
trans-4-(Aminomethyl)cyclohexanecarboxylic Acid Methyl Ester Hydrochloride
methyl 4-(aminomethyl)cyclohexane-1-carboxylate;hydrochloride

Specifications

Undesignated
29275-88-5
MFCD00797465
PZPSRTWFJCGJLP-UHFFFAOYSA-N
methyl 4-(aminomethyl)cyclohexane-1-carboxylate;hydrochloride
12721445
Crystal-Powder at 20°C
Methyl trans-4-(Aminomethyl)cyclohexanecarboxylate Hydrochloride
5g
C9H18ClNO2
trans-4-(Aminomethyl)cyclohexanecarboxylic Acid Methyl Ester Hydrochloride
COC(=O)C1CCC(CC1)CN.Cl
207.698
207.70
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No