Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl trans-3-nonenoate, 98%

$26.31 - $99.59

Chemical Identifiers

Chemical Name or Material Methyl trans-3-nonenoate
CAS 36781-67-6
Molecular Formula CH3(CH2)4CH=CHCH2CO2CH3
Formula Weight 170.25
MDL Number MFCD00065419
Synonym trans-Nonenoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1928814

  SDS
alfa aesar™
A1928814
25g Each for $99.59

AAA1928806

  SDS
alfa aesar™
A1928806
5g Each for $26.31
Description & Specifications

Specifications

Formula Weight 170.25
Chemical Name or Material Methyl trans-3-nonenoate
Assay 98%
Molecular Formula CH3(CH2)4CH=CHCH2CO2CH3
MDL Number MFCD00065419
CAS 36781-67-6
Synonym trans-Nonenoic acid methyl ester
Quantity 25g