missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl trans-3-(benzyloxycarbonylamino)cyclohexanecarboxylate, 97%

$68.31 - $990.04

Chemical Identifiers

CAS 1035325-22-4
Molecular Formula C16H21NO4
Molecular Weight (g/mol) 291.347
MDL Number MFCD21365071
InChI Key OGLDGYYJEHFVKN-KGLIPLIRSA-N
Synonym trans-methyl-3-benzyloxy carbonyl-amino cyclohexanecarboxylate
PubChem CID 118309547
IUPAC Name methyl (1R,3S)-3-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylate
SMILES COC(=O)C1CCCC(C1)NC(=O)OCC2=CC=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62560MD View Documents Alfa Aesar™
H62560MD
250mg Each for $68.31
 
AAH6256003 View Documents Alfa Aesar™
H6256003
1g Each for $217.43
 
AAH6256006 View Documents Alfa Aesar™
H6256006
5g Each for $990.04
 
Description

Description

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

1035325-22-4
291.347
OGLDGYYJEHFVKN-KGLIPLIRSA-N
118309547
COC(=O)C1CCCC(C1)NC(=O)OCC2=CC=CC=C2
C16H21NO4
MFCD21365071
trans-methyl-3-benzyloxy carbonyl-amino cyclohexanecarboxylate
methyl (1R,3S)-3-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylate

Specifications

250mg
C16H21NO4
trans-methyl-3-benzyloxy carbonyl-amino cyclohexanecarboxylate
COC(=O)C1CCCC(C1)NC(=O)OCC2=CC=CC=C2
291.347
291.34
Methyl trans-3-(benzyloxycarbonylamino)cyclohexanecarboxylate
1035325-22-4
MFCD21365071
OGLDGYYJEHFVKN-KGLIPLIRSA-N
methyl (1R,3S)-3-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylate
118309547
97%