Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl trans-2-octenoate, 90+%, remainder mainly cis-isomer

$44.95 - $158.54

Chemical Identifiers

Chemical Name or Material Methyl trans-2-octenoate
CAS 7367-81-9
Molecular Formula C9H16O2
Formula Weight 156.22
MDL Number MFCD00036620
Synonym trans-2-Octenoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1961206

  SDS
alfa aesar™
A1961206
5g Each for $44.95

AAA1961214

  SDS
alfa aesar™
A1961214
25g Each for $158.54
Description & Specifications

Specifications

Formula Weight 156.22
Chemical Name or Material Methyl trans-2-octenoate
Assay ≥90%
Assay Percent Notes remainder mainly cis-isomer
Molecular Formula C9H16O2
MDL Number MFCD00036620
CAS 7367-81-9
Synonym trans-2-Octenoic acid methyl ester
Health Hazard 1 Warning
Boiling Point 70°C (4mmHg)
Quantity 5g
Flash Point 79°C (174°F)
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.