Learn More
Methyl tiglate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1196418
Description
It is employed as a perfuming agent in cosmetics industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a perfuming agent in cosmetics industry.
Solubility
Soluble in alcohol, water 3813 mg/L @ 25°C (est).
Notes
Store away from oxidizing agents. Keep the container tightly closed. Store in cool, dry conditions in well ventilated place.
Specifications
Methyl tiglate | |
0.95 | |
39°C (102°F) | |
1.437 | |
50 g | |
1720455 | |
methyl tiglate, tiglic acid methyl ester, methyl 2-methylbut-2-enoate, methyl 2-methyl-2-butenoate, methyl e-2-methylcrotonate, methyl 2-methylcrotonate, methyl alpha-methylcrotonate, methyl trans-2-methylcrotonate, 2-butenoic acid, 2-methyl-, methyl ester, e, crotonic acid, 2-methyl-, methyl ester, e | |
YYJWBYNQJLBIGS-PLNGDYQASA-N | |
methyl (E)-2-methylbut-2-enoate | |
5323652 | |
98% |
6622-76-0 | |
138°C to 139°C | |
C6H10O2 | |
MFCD00016654 | |
UN3272 | |
14,9433 | |
Soluble in alcohol,water 3813mg/L @ 25°C (est). | |
COC(=O)C(\C)=C/C | |
114.14 | |
114.15 |
Chemical Identifiers
6622-76-0 | |
114.14 | |
YYJWBYNQJLBIGS-PLNGDYQASA-N | |
5323652 | |
COC(=O)C(\C)=C/C |
C6H10O2 | |
MFCD00016654 | |
methyl tiglate, tiglic acid methyl ester, methyl 2-methylbut-2-enoate, methyl 2-methyl-2-butenoate, methyl e-2-methylcrotonate, methyl 2-methylcrotonate, methyl alpha-methylcrotonate, methyl trans-2-methylcrotonate, 2-butenoic acid, 2-methyl-, methyl ester, e, crotonic acid, 2-methyl-, methyl ester, e | |
methyl (E)-2-methylbut-2-enoate |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 229-575-8
RTECSNumber : EM9254600
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only