Please login to your online account to display your discounted pricing

Methyl tetrahydrothiopyran-4-carboxylate, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC61922DE

 View more versions of this product

Catalog No. CC61922DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 160.24
Quantity 5g
Assay 97%
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 128094-82-6
Chemical Name or Material Methyl tetrahydrothiopyran-4-carboxylate
Molecular Formula C7H12O2S

Specifications

Chemical Name or Material Methyl tetrahydrothiopyran-4-carboxylate
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H12O2S
Formula Weight 160.24