Please login to your online account to display your discounted pricing

Methyl tetrahydrothiopyran-4-carboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl tetrahydrothiopyran-4-carboxylate
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H12O2S
Formula Weight 160.24
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC61922DE

 
fisher scientific company
CC61922DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC61922DA

 
fisher scientific company
CC61922DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC61922EA

 
fisher scientific company
CC61922EA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 160.24
Quantity 5g
Assay 97%
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 128094-82-6
Chemical Name or Material Methyl tetrahydrothiopyran-4-carboxylate
Molecular Formula C7H12O2S