Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl tert-butylacetate, 98%

Chemical Identifiers

Chemical Name or Material Methyl tert-butylacetate
CAS 10250-48-3
Molecular Formula (CH3)3CCH2COOCH3
Formula Weight 130.19
MDL Number MFCD00043646
UN Number UN3272
Beilstein 1747895
Synonym tert-Butylacetic acid methyl ester; Methyl 3,3-dimethylbutyrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0700509

  SDS
alfa aesar™
L0700509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 130.19
Chemical Name or Material Methyl tert-butylacetate
Molecular Formula (CH3)3CCH2COOCH3
MDL Number MFCD00043646
CAS 10250-48-3
Boiling Point 126° to 128°C
Flash Point 24°C (75°F)
Assay 98%
UN Number UN3272
Beilstein 1747895
Synonym tert-Butylacetic acid methyl ester; Methyl 3,3-dimethylbutyrate
Health Hazard 1 Warning
Quantity 10g
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

Applications
Methyl tert-butylacetate is an oxygenated solvent used alone or in blends for coatings, industrial cleaning and surface treatment. It is used as an intermediate to manufacture pharmaceuticals, synthetic flavorings and other organic compounds.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.