Please login to your online account to display your discounted pricing

Methyl sulfoxide-d6, for NMR, 99.5+% atom D, ACROS Organics™


Quantity 1L

Manufacturer: thermo fisher scientific  321290010

 View more versions of this product

Catalog No. AC321290010

  • / Each
 

Description & Specifications

Chemical Identifiers

Additional Information Vapor Pressure: 2.5mbar at 20°C
Deuterium (D) 99.50% min.
Water 0.05% max. (Coulometric)
Density 1.1900g/mL
Decomposition Information >190°C
Quantity 1L
Color Clear colorless
Chemical Name or Material Methyl sulfoxide-d6
Name Note 99.5+ atom % D
CAS 2206-27-1
Purity Grade For NMR
Physical Form Clear colorless liquid
Molecular Formula C2D6OS
Linear Formula (CD3)2SO
Formula Weight 84.18g/mol
MDL Number MFCD00002090
Beilstein 01, IV, 1279
Fieser 13,124; 16,177
Synonym Dimethyl-d6 sulfoxide
Health Hazard 1 HYGROSCOPIC
EINECS Number 218-617-0
TSCA TSCA

Specifications

Chemical Name or Material Methyl sulfoxide-d6
Name Note 99.5+ atom % D
CAS 2206-27-1
Purity Grade For NMR
Molecular Formula C2D6OS
Linear Formula (CD3)2SO
Formula Weight 84.18g/mol
MDL Number MFCD00002090
Synonym Dimethyl-d6 sulfoxide

For NMR

Accessories

HYGROSCOPIC

SDS & Safety Documents