Please login to your online account to display your discounted pricing

Methyl sulfoxide-d6, for NMR, 100.0% atom D, min. 99.96% atom D, ACROS Organics™

$178.79 - $616.68

Chemical Identifiers

Chemical Name or Material Methyl sulfoxide-d6
CAS 2206-27-1
Purity Grade For NMR
Molecular Formula C2D6OS
Linear Formula (CD3)2SO
Formula Weight 84.18
MDL Number MFCD00002090
Synonym Dimethyl-d6 sulfoxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC169020050

  SDS
thermo fisher scientific
169020050
5mL Each for $178.79

AC169020250

  SDS
thermo fisher scientific
169020250
25mL Each for $616.68
Description & Specifications

Specifications

Chemical Name or Material Methyl sulfoxide-d6
CAS 2206-27-1
Deuterium (D) 99.95% min.
Health Hazard 1 HYGROSCOPIC
EINECS Number 218-617-0
TSCA TSCA
Water 0.02% max. (Coulometric)
Density 1.1900g/mL
Purity Grade For NMR
Physical Form Liquid
Quantity 5mL
Color Clear colorless
Molecular Formula C2D6OS
Linear Formula (CD3)2SO
Formula Weight 84.18
MDL Number MFCD00002090
Beilstein 01, IV, 1279
Fieser 13,124; 16,177
Synonym Dimethyl-d6 sulfoxide

For NMR

Safety and Handling

HYGROSCOPIC