Please login to your online account to display your discounted pricing

Methyl sulfoxide-d6, for NMR, with 0.03% TMS, 99.9 atom% D, ACROS Organics™

$59.05 - $132.87

Chemical Identifiers

Chemical Name or Material Methyl sulfoxide-d6
Name Note with 0.03% TMS, 99.9 atom%D
CAS 2206-27-1
Purity Grade For NMR
Molecular Formula C2D6OS
Formula Weight 84.18
MDL Number MFCD00002090
Synonym Dimethyl-d6 sulfoxide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC351450100

  SDS
thermo fisher scientific
351450100
Glass bottle 10mL Each for $59.05

AC351450250

  SDS
thermo fisher scientific
351450250
Glass bottle 25mL Each for $132.87
Description & Specifications

Specifications

Formula Weight 84.18
Physical Form Liquid
Quantity 10mL
Color Clear colorless
Beilstein 01, IV, 1279
Fieser 13,124; 16,177
Purity Grade For NMR
Density 1.1900g/mL
Molecular Formula C2D6OS
MDL Number MFCD00002090
CAS 2206-27-1
Chemical Name or Material Methyl sulfoxide-d6
Deuterium (D) 99.85% min.
Water 0.03% max. (Coulometric)
Synonym Dimethyl-d6 sulfoxide
Packaging Glass bottle
Name Note with 0.03% TMS, 99.9 atom%D

For NMR