missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl sulfoxide, 99.7%, Extra Dry, AcroSeal™, ACROS Organics™
Manufacturer: Fisher Scientific Acros 610420010
Specifications
Methyl sulfoxide | |
Vapor Pressure: 0.55mbar at 20°C | |
C2H6OS | |
MFCD00002089 | |
01,296; 02,157; 03,119; 04,192; 05,263; 06,225; 07,133; 08,198; 09,189; 10,166; 12,212; 13,124; 15,146; 16,149; 17,127 | |
demasorb, demavet, demsodrox, dimethyl sulfoxide, dimethyl sulphoxide, dimethylsulfoxide, dimexide, dmso, methane, sulfinylbis, methyl sulfoxide | |
CS(=O)C | |
78.129 | |
CHEBI:28262 | |
≥99.7% | |
2.7 (Air = 1.0) | |
≤50ppm | |
1L |
67-68-5 | |
99.7% | |
(CH3)2SO | |
01, IV, 1277 | |
15, 3285 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
methylsulfinylmethane | |
679 | |
78.13 | |
Extra Dry | |
Pass Test | |
>190°C |
Chemical Identifiers
67-68-5 | |
78.129 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
679 | |
methylsulfinylmethane |
C2H6OS | |
MFCD00002089 | |
demasorb, demavet, demsodrox, dimethyl sulfoxide, dimethyl sulphoxide, dimethylsulfoxide, dimexide, dmso, methane, sulfinylbis, methyl sulfoxide | |
CHEBI:28262 | |
CS(=O)C |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 200-664-3
RTECSNumber : PV6210000
TSCA : TSCA