missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Sulfide 99.0+%, TCI America™
Supplier: TCI America M0431100ML
Specifications
| Methyl Sulfide | |
| -98°C | |
| 37°C | |
| MFCD00008562 | |
| 1164 | |
| QMMFVYPAHWMCMS-UHFFFAOYSA-N | |
| (methylsulfanyl)methane | |
| 1068 | |
| 62.13 | |
| Liquid |
| 75-18-3 | |
| Colorless | |
| C2H6S | |
| 100 mL | |
| dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane | |
| CSC | |
| 62.13 | |
| CHEBI:17437 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 75-18-3 | |
| 62.13 | |
| QMMFVYPAHWMCMS-UHFFFAOYSA-N | |
| 1068 | |
| (methylsulfanyl)methane |
| C2H6S | |
| MFCD00008562 | |
| dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane | |
| CHEBI:17437 | |
| CSC |
Safety and Handling
EINECSNumber : (2)-0466
RTECSNumber : PV5075000
TSCA : Yes