Learn More
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 127760025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl sulfide | |
-98.0°C | |
0.8460g/mL | |
−48°C | |
0.2% max. Methylmercaptan (GC) | |
C2H6S | |
(CH3)2S | |
2.5 L | |
02,156; 04,190; 05,260; 15,88; 16,84; 17,198 | |
15, 3281 | |
Solubility in water: 2g/L (20°C). Other solubilities: soluble in alcohol and ether | |
CSC | |
62.13 | |
CHEBI:17437 | |
99+% | |
Liquid |
75-18-3 | |
Colorless | |
38.0°C | |
Authentic | |
Glass bottle | |
1.4351 | |
MFCD00008562 | |
01, 288 | |
0.846 | |
dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane | |
QMMFVYPAHWMCMS-UHFFFAOYSA-N | |
methylsulfanylmethane | |
1068 | |
62.13 | |
Extra Pure |
Chemical Identifiers
75-18-3 | |
62.13 | |
QMMFVYPAHWMCMS-UHFFFAOYSA-N | |
1068 | |
methylsulfanylmethane |
C2H6S | |
MFCD00008562 | |
dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane | |
CHEBI:17437 | |
CSC |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g
GHS Signal Word: Danger
EINECSNumber : 200-846-2
RUO – Research Use Only