missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Salicylate, NF, 98-102%, Spectrum™ Chemical

ME170, 119-36-8, C8H8O3

Supplier:  Spectrum Chemical Mfg Cor ME1704LTGL

 View more versions of this product

Catalog No. 18604455



Description

Description

Spectrum™ Chemical Methyl Salicylate, NF is used to treat minor aches and pains of the muscles and joints. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

119-36-8
Amber Glass Bottle
4 L
COC(=O)C1=CC=CC=C1O
152.15
NF
100%
C8H8O3
OSWPMRLSEDHDFF-UHFFFAOYSA-N
methyl 2-hydroxybenzoate
98 to 102%

Chemical Identifiers

119-36-8
152.15
methyl 2-hydroxybenzoate
C8H8O3
OSWPMRLSEDHDFF-UHFFFAOYSA-N
COC(=O)C1=CC=CC=C1O
SDS
Documents

Documents

Promotions

Promotions