missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl salicylate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 220490025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl salicylate | |
-8°C to 7°C | |
1.1825g/mL | |
110°C | |
98.5% min. (GC) | |
C8H8O3 | |
2-(HO)C6H4CO2CH3 | |
2.5 L | |
1.1825 | |
methyl salicylate, wintergreen oil, gaultheria oil, betula oil, teaberry oil, sweet birch oil, oil of wintergreen, analgit, spicewood oil, 2-hydroxybenzoic acid methyl ester | |
OSWPMRLSEDHDFF-UHFFFAOYSA-N | |
methyl 2-hydroxybenzoate | |
4133 | |
152.15 | |
Liquid |
119-36-8 | |
Colorless to Yellow | |
222.0°C | |
Authentic | |
Glass bottle | |
1.5350 to 1.5380 | |
MFCD00002214 | |
10,7 | |
15,6193 | |
Solubility in water: 0.07 % (20°C). Other solubilities: soluble in glacial acetic acid, soluble in ether and chloroform, in ethanol 80 vol. %:1:2 v/v (20°C), diethyl phthalate:1:1 v/v (20°C), 1,2-propanediol:1/7 v/v (20°C) | |
COC(=O)C1=CC=CC=C1O | |
152.15 | |
CHEBI:31832 | |
99% |
Chemical Identifiers
119-36-8 | |
152.15 | |
OSWPMRLSEDHDFF-UHFFFAOYSA-N | |
4133 | |
methyl 2-hydroxybenzoate |
C8H8O3 | |
MFCD00002214 | |
methyl salicylate, wintergreen oil, gaultheria oil, betula oil, teaberry oil, sweet birch oil, oil of wintergreen, analgit, spicewood oil, 2-hydroxybenzoic acid methyl ester | |
CHEBI:31832 | |
COC(=O)C1=CC=CC=C1O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 204-317-7
RUO – Research Use Only