Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl (S)-(+)-mandelate, 99%


Quantity 5g

Manufacturer: alfa aesar™  L1407006

 View more versions of this product

Catalog No. AAL1407006

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 166.18
Chemical Name or Material Methyl (S)-(+)-mandelate
Assay 99%
Molecular Formula C9H10O3
MDL Number MFCD00064246
CAS 21210-43-5
Beilstein 2614741
Synonym (S)-(+)-alpha-Hydroxyphenylacetic acid methyl ester; (S)-(+)-Mandelic acid methyl ester
Melting Point 56° to 58°C
Quantity 5g
Flash Point >110°C (230°F)
Optical Rotation +144° (c= 1 in Methanol)

Specifications

Chemical Name or Material Methyl (S)-(+)-mandelate
CAS 21210-43-5
Molecular Formula C9H10O3
Formula Weight 166.18
MDL Number MFCD00064246
Beilstein 2614741
Synonym (S)-(+)-alpha-Hydroxyphenylacetic acid methyl ester; (S)-(+)-Mandelic acid methyl ester