Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl (S)-(+)-mandelate, 99%

$127.91 - $127.91

Chemical Identifiers

Chemical Name or Material Methyl (S)-(+)-mandelate
CAS 21210-43-5
Molecular Formula C9H10O3
Formula Weight 166.18
MDL Number MFCD00064246
Beilstein 2614741
Synonym (S)-(+)-alpha-Hydroxyphenylacetic acid methyl ester; (S)-(+)-Mandelic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1407006

  SDS
alfa aesar™
L1407006
5g Each for $127.91
Description & Specifications

Specifications

Formula Weight 166.18
Chemical Name or Material Methyl (S)-(+)-mandelate
Assay 99%
Molecular Formula C9H10O3
MDL Number MFCD00064246
CAS 21210-43-5
Beilstein 2614741
Synonym (S)-(+)-alpha-Hydroxyphenylacetic acid methyl ester; (S)-(+)-Mandelic acid methyl ester
Melting Point 56° to 58°C
Quantity 5g
Flash Point >110°C (230°F)
Optical Rotation +144° (c= 1 in Methanol)