Please login to your online account to display your discounted pricing

Methyl (S)-(+)-mandelate, ≥98%, Acros Organics


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  264680050

 View more versions of this product

Catalog No. AC264680050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Crystalline Powder
Formula Weight 166.18
Quantity 5g
Color White
Beilstein 10, 114
Fieser 16,219
TSCA TSCA
Specific Rotation Condition + 142.00 (20.00°C c=2,methanol)
Melting Point 54° to 58°C
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material Methyl (S)-(+)-mandelate
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98+%
Specific Rotation +142° to +146° (20°C, 589nm) (c=2, CH3OH)
Assay Percent Range 98+%
Molecular Formula C9H10O3
Linear Formula C6H5CH(OH)CO2CH3
MDL Number MFCD00064246
CAS 21210-43-5

Specifications

Chemical Name or Material Methyl (S)-(+)-mandelate
Name Note 98+%
CAS 21210-43-5
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C9H10O3
Linear Formula C6H5CH(OH)CO2CH3
Formula Weight 166.18
MDL Number MFCD00064246