Please login to your online account to display your discounted pricing

Methyl (S)-(+)-mandelate, ≥98%, Acros Organics

Chemical Identifiers

Chemical Name or Material Methyl (S)-(+)-mandelate
Name Note 98+%
CAS 21210-43-5
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C9H10O3
Linear Formula C6H5CH(OH)CO2CH3
Formula Weight 166.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC264680010

  SDS
thermo fisher scientific
264680010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC264680050

  SDS
thermo fisher scientific
264680050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 166.18
Physical Form Crystalline Powder
Assay ≥98.0%
Quantity 1g
Color White
TSCA TSCA
Melting Point 54° to 58°C
Specific Rotation Condition + 142.00 (20.00°C c=2,methanol)
Beilstein 10, 114
Fieser 16,219
CAS Max % 100.0
CAS Min % 98.0
Specific Rotation +142° to +146° (20°C, 589nm) (c=2, CH3OH)
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C9H10O3
Chemical Name or Material Methyl (S)-(+)-mandelate
Name Note 98+%
MDL Number MFCD00064246
CAS 21210-43-5
Assay Percent Range 98+%
Linear Formula C6H5CH(OH)CO2CH3