Please login to your online account to display your discounted pricing

Methyl (S)-(-)3-BOC-2,2-dimethyl-4-oxazolidinecarboxylate, 93%, Tech., ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl (S)-(-)3-BOC-2, 2-dimethyl-4-oxazolidinecarboxylate
Name Note tech., 93%
CAS 108149-60-6
Purity Grade Technical Grade
Assay ≥91%
Assay Percent Range 93%
Molecular Formula C12H21NO5
Formula Weight 259.3
MDL Number MFCD00192279
Synonym 2, 2-Dimethyl oxazolidine-3, 4-dicarboxylic acid 3-tert-butyl ester 4-methyl ester
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC301260010

  SDS
thermo fisher scientific
AC301260010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥91%
Physical Form Liquid
Purity Grade Technical Grade
Formula Weight 259.3
Quantity 1g
Color Clear yellow
Specific Rotation Condition −53.5° (20°C c=1,CHCl3)
Density 1.082g/mL
Infrared Spectrum Authentic
Assay Percent Range 93%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00192279
Synonym 2, 2-Dimethyl oxazolidine-3, 4-dicarboxylic acid 3-tert-butyl ester 4-methyl ester
Refractive Index 1.4420 to 1.4440 (20°C, 589nm)
Packaging Glass bottle
Specific Rotation -50° to -57° (20°C, 589nm) (c=1, CHCl3)
Chemical Name or Material Methyl (S)-(-)3-BOC-2, 2-dimethyl-4-oxazolidinecarboxylate
Molecular Formula C12H21NO5
Name Note tech., 93%
CAS 108149-60-6
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.