missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl (S)-(+)-2-oxopyrrolidine-5-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H6254314

 View more versions of this product

Catalog No. AAH6254314



Specifications

Specifications

Methyl (S)-(+)-2-oxopyrrolidine-5-carboxylate
1.4860
Yellow
4931-66-2
MFCD00080931
HQGPKMSGXAUKHT-BYPYZUCNSA-N
methyl (2S)-5-oxopyrrolidine-2-carboxylate
78646
Liquid
1.226g/mL
90°C (0.3mmHg)
25g
C6H9NO3
h-pyr-ome
COC(=O)C1CCC(=O)N1
143.142
143.14
97%

Chemical Identifiers

4931-66-2
143.142
HQGPKMSGXAUKHT-BYPYZUCNSA-N
78646
COC(=O)C1CCC(=O)N1
C6H9NO3
MFCD00080931
h-pyr-ome
methyl (2S)-5-oxopyrrolidine-2-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents