missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl (S)-(-)-2-Isocyanato-3-methylbutyrate, TCI America™

$105.42 - $328.48

Chemical Identifiers

CAS 30293-86-8
Molecular Formula C7H11NO3
Molecular Weight (g/mol) 157.169
MDL Number MFCD00191531
InChI Key JCVZXXSBCISMLJ-LURJTMIESA-N
Synonym (S)-(-)-2-Isocyanato-3-methylbutyric Acid Methyl Ester, Methyl (S)-(-)-2-Isocyanatoisovalerate
PubChem CID 10397066
IUPAC Name methyl (2S)-2-isocyanato-3-methylbutanoate
SMILES CC(C)C(C(=O)OC)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I04661G View Documents TCI America
I04661G
1g Each for $105.42
 
I04665G View Documents TCI America
I04665G
5g Each for $328.48
 
Specifications

Chemical Identifiers

30293-86-8
157.169
JCVZXXSBCISMLJ-LURJTMIESA-N
10397066
CC(C)C(C(=O)OC)N=C=O
C7H11NO3
MFCD00191531
(S)-(-)-2-Isocyanato-3-methylbutyric Acid Methyl Ester, Methyl (S)-(-)-2-Isocyanatoisovalerate
methyl (2S)-2-isocyanato-3-methylbutanoate

Specifications

76°C
1g
C7H11NO3
(S)-(-)-2-Isocyanato-3-methylbutyric Acid Methyl Ester, Methyl (S)-(-)-2-Isocyanatoisovalerate
CC(C)C(C(=O)OC)N=C=O
157.169
157.17
≥98.0% (GC)
Yellow
30293-86-8
MFCD00191531
JCVZXXSBCISMLJ-LURJTMIESA-N
methyl (2S)-2-isocyanato-3-methylbutanoate
10397066
Clear Liquid at 20°C
Methyl (S)-(-)-2-Isocyanato-3-methylbutyrate
Safety and Handling

Safety and Handling

TSCA : No