missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl Red Solution R, Ricca Chemical
$146.72 - $146.72
Chemical Identifiers
CAS | 493-52-7 |
---|---|
Molecular Formula | C15H15N3O2 |
Molecular Weight (g/mol) | 269.30 |
InChI Key | CEQFOVLGLXCDCX-UHFFFAOYSA-N |
PubChem CID | 10303 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
5047-4
|
Ricca Chemical Company
50474 |
120 mL | Amber Glass Bottle |
Each for $146.72
|
|
||||
Chemical Identifiers
493-52-7 | |
269.30 | |
10303 |
C15H15N3O2 | |
CEQFOVLGLXCDCX-UHFFFAOYSA-N |
Specifications
493-52-7,64-17-5,493-52-7,1310-73-2,7732-18-5 | |
58.0,44.39,0.06,0.02 | |
CEQFOVLGLXCDCX-UHFFFAOYSA-N | |
10303 | |
Red | |
Liquid |
55.0,42.65,0.05,0.02 | |
C15H15N3O2 | |
269.30 | |
Amber Glass Bottle | |
120 mL | |
Methyl Red Solution, Reagent |