Please login to your online account to display your discounted pricing

Methyl Red Sodium Salt, ACS Grade, LabChem™

$130.06 - $130.06

Chemical Identifiers

Chemical Name or Material Methyl Red Sodium Salt
CAS 845-10-3
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C15H14N3O2Na
Formula Weight 291.28
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC170807

 
lab chem
LC170807
10g Each for $130.06
Description & Specifications

Specifications

Formula Weight 291.28
Chemical Name or Material Methyl Red Sodium Salt
Quantity 10g
Purity Grade ACS Grade
Physical Form Powder
Linear Formula C15H14N3O2Na
CAS 845-10-3
Identification Passes Test
Packaging Amber Glass
CAS Max % 100
Health Hazard 2 GHS H Statement
Product is not hazardous.
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Solubility Soluble in water
Recommended Storage Room Temperature
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Color Reddish brown
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.

Room Temperature