missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl Red - Methylene Blue Solution (Ethanol Soln.) [mixed Indicator], TCI America™
$61.11 - $180.28
Chemical Identifiers
MDL Number | MFCD00079686 |
---|---|
Synonym | Methylene Blue - Methyl Red Solution |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
M0161100ML |
TCI America
M0161100ML |
100mL |
Each for $61.11
|
|
M0161500ML |
TCI America
M0161500ML |
500mL |
Each for $180.28
|
|
Chemical Identifiers
MFCD00079686 | |
Methylene Blue - Methyl Red Solution |
Specifications
MFCD00079686 | |
Methylene Blue - Methyl Red Solution | |
Green | |
Methyl Red - Methylene Blue Solution (Ethanol Soln.) [mixed Indicator] |
1170 | |
Liquid at 20°C | |
100mL |
Safety and Handling
TSCA : No