Please login to your online account to display your discounted pricing

Methyl Red-Methylene Blue, Mixed Indicator, for Nitrogen (Ammonia), Nitrogen (Organic), Certified, LabChem™


Quantity 500mL
Packaging Poly Bottle

Manufacturer: lab chem  LC171701

 View more versions of this product

Catalog No. LC171701

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Formula Weight 291.28
Chemical Name or Material Methyl Red-Methylene Blue
Quantity 500mL
Flash Point 13°C
CAS Max % 4.96
CAS 845-10-3
CAS Max % 0.13
CAS 61-73-4
CAS Max % 0.07
Name Note Mixed Indicator, for Nitrogen (Ammonia), Nitrogen (Organic)
Physical Form Liquid
Linear Formula C15H14N3O2Na
CAS 64-17-5
UN Number UN1170
Identification Passes Test
Health Hazard 1 Danger
Packaging Poly Bottle
CAS Max % 89.88
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Suspected of damaging the unborn child (oral).
Causes damage to organs (central nervous system, kidneys, liver, optic nerve).
CAS 67-63-0
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
In case of fire: Use alcohol resistant foam, carbon dioxide (CO2), extinguishing powder to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Solubility Soluble in water
CAS Max % 4.96
Recommended Storage Room Temperature
Decomposition Information Carbon monoxide; Carbon dioxide; May release flammable gases
CAS 67-56-1
Boiling Point 78°C
Melting Point −115°C
Color Dark, green/black

Specifications

Chemical Name or Material Methyl Red-Methylene Blue
CAS 64-17-5
CAS Max % 89.88
CAS 67-63-0
CAS Max % 4.96
CAS 67-56-1
CAS Max % 4.96
CAS 845-10-3
CAS Max % 0.13
CAS 61-73-4
CAS Max % 0.07
Name Note Mixed Indicator, for Nitrogen (Ammonia), Nitrogen (Organic)
Purity Grade Certified Grade
Linear Formula C15H14N3O2Na
Formula Weight 291.28
UN Number UN1170
Accessories

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
In case of fire: Use alcohol resistant foam, carbon dioxide (CO2), extinguishing powder to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Highly flammable liquid and vapor.
Suspected of damaging the unborn child (oral).
Causes damage to organs (central nervous system, kidneys, liver, optic nerve).

Room Temperature