Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl Red, 0.1% w/v solution in ethanol

Chemical Identifiers

Chemical Name or Material Methyl Red
Formula Weight 269.3
MDL Number MFCD00002425
UN Number UN1170
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38698AE

  SDS
alfa aesar™
38698AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA38698AP

  SDS
alfa aesar™
38698AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 269.3
Chemical Name or Material Methyl Red
Quantity 100mL
TSCA TSCA
Concentration 0.1% w/v solution in ethanol
Physical Form Liquid
MDL Number MFCD00002425
UN Number UN1170
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H224
Extremely flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H224
Extremely flammable liquid and vapor.