missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl Red, 0.02% in Methanol, pH 4.2 to 6.3 Pink to Yellow, Certified, LabChem™
Supplier: LabChem LC171002
Specifications
Methyl Red | |
0.02% in Methanol, pH 4.2 to 6.3 Pink to Yellow | |
99.98,0.02 | |
CH4O | |
MFCD00004595 | |
Passes Test | |
CO | |
32.04 | |
Certified | |
0.79g/mL | |
0.79g/mL | |
1 L | |
Liquid |
-98°C | |
65°C | |
845-10-3 | |
C15H14N3O2Na | |
Soluble in water | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
291.28 | |
Poly Bottle | |
Carbon monoxide; Carbon dioxide; May release flammable gases | |
Orange | |
9.7°C |