Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl (R)-(-)-mandelate, 99%

$115.57 - $115.57

Chemical Identifiers

Chemical Name or Material Methyl (R)-(-)-mandelate
CAS 20698-91-3
Molecular Formula C9H10O3
Formula Weight 166.18
MDL Number MFCD00064247
Beilstein 3589447
Synonym (R)-(-)-alpha-Hydroxyphenylacetic acid methyl ester; (R)-(-)-Mandelic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1406506

  SDS
alfa aesar™
L1406506
5g Each for $115.57
Description & Specifications

Specifications

Formula Weight 166.18
Chemical Name or Material Methyl (R)-(-)-mandelate
CAS 20698-91-3
MDL Number MFCD00064247
Melting Point 56° to 58°C
Quantity 5g
Optical Rotation −144° (c=1 in Methanol)
Flash Point >110°C (230°F)
Molecular Formula C9H10O3
Synonym (R)-(-)-alpha-Hydroxyphenylacetic acid methyl ester; (R)-(-)-Mandelic acid methyl ester
Assay 99%
Beilstein 3589447