Please login to your online account to display your discounted pricing

Methyl (R)-(+)-lactate, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC414490010

 View more versions of this product

Catalog No. AC414490010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 104.11
Quantity 1g
Color Clear colorless
Specific Rotation Condition + 8.50 (20.00°C c=neat)
Density 1.0900g/mL
UN Number 3272
Chemical Name or Material Methyl (R)-(+)-lactate
Refractive Index 1.4120 to 1.4140 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C4H8O3
MDL Number MFCD00004517
Health Hazard 1 Warning
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Rotation +8° to +9° (20°C, 589nm) (neat)
CAS 17392-83-5
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Methyl (R)-(+)-lactate
CAS 17392-83-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H8O3
Formula Weight 104.11
MDL Number MFCD00004517
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

SDS & Safety Documents