missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl (R)-(-)-3-Hydroxybutyrate 98.0+%, TCI America™
Supplier: TCI America H070525ML
Specifications
| Methyl (R)-(-)-3-Hydroxybutyrate | |
| Yellow | |
| C5H10O3 | |
| 25 mL | |
| LDLDJEAVRNAEBW-SCSAIBSYSA-N | |
| methyl (3R)-3-hydroxybutanoate | |
| 2724279 | |
| ≥98.0% (GC) |
| 3976-69-0 | |
| 75°C | |
| MFCD00063289 | |
| methyl r---3-hydroxybutyrate, r-methyl 3-hydroxybutanoate, methyl 3r-3-hydroxybutanoate, methyl r-3-hydroxybutyrate, r-3-hydroxybutyric acid methyl ester, methyl r-3-hydroxybutanoate, r---3-hydroxybutyric acid methyl ester, r---methyl 3-hydroxybutyrate, r---3-hydroxy-n-butyric acid methyl ester | |
| CC(CC(=O)OC)O | |
| 118.132 | |
| 118.13 | |
| Liquid |
Chemical Identifiers
| 3976-69-0 | |
| 118.132 | |
| LDLDJEAVRNAEBW-SCSAIBSYSA-N | |
| 2724279 | |
| CC(CC(=O)OC)O |
| C5H10O3 | |
| MFCD00063289 | |
| methyl r---3-hydroxybutyrate, r-methyl 3-hydroxybutanoate, methyl 3r-3-hydroxybutanoate, methyl r-3-hydroxybutyrate, r-3-hydroxybutyric acid methyl ester, methyl r-3-hydroxybutanoate, r---3-hydroxybutyric acid methyl ester, r---methyl 3-hydroxybutyrate, r---3-hydroxy-n-butyric acid methyl ester | |
| methyl (3R)-3-hydroxybutanoate |
Safety and Handling
EINECSNumber : (2)-1430
RTECSNumber : ET4700000
TSCA : No