missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl (R)-(+)-3-BOC-2,2-dimethyl-4-oxazolidinecarboxylate, 95%, ACROS Organics™
Chemical Identifiers
CAS | 95715-86-9 |
---|---|
Molecular Formula | C12H21NO5 |
Molecular Weight (g/mol) | 259.3 |
MDL Number | MFCD00674041 |
InChI Key | ZNBUXTFASGDVCL-MRVPVSSYSA-N |
Synonym | 3-tert-butyl 4-methyl 4r-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate, 3-tert-butyl 4-methyl r-+-2,2-dimethyloxazolidine-3,4-dicarboxylate, methyl 3-tert-butoxycarbonyl-2,2-dimethyloxazolidine-4-r-carboxylate, methyl r-+-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate, methyl r-+-3-tert-butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylate, methyl r-n-boc-2,2-dimethyloxazolidine-4-carboxylate, r-+-3-boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, r-+-3-tert-butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylic acid methyl ester, r---3-boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, r-3-tert-butyl 4-methyl 2,2-dimethyloxazolidine-3,4-dicarboxylate |
PubChem CID | 688220 |
IUPAC Name | 3-O-tert-butyl 4-O-methyl (4R)-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate |
SMILES | CC1(N(C(CO1)C(=O)OC)C(=O)OC(C)(C)C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC426350010
|
Thermo Fisher Scientific
AC426350010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC426350050
|
Thermo Fisher Scientific
AC426350050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
95715-86-9 | |
259.3 | |
ZNBUXTFASGDVCL-MRVPVSSYSA-N | |
688220 | |
CC1(N(C(CO1)C(=O)OC)C(=O)OC(C)(C)C)C |
C12H21NO5 | |
MFCD00674041 | |
3-tert-butyl 4-methyl 4r-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate, 3-tert-butyl 4-methyl r-+-2,2-dimethyloxazolidine-3,4-dicarboxylate, methyl 3-tert-butoxycarbonyl-2,2-dimethyloxazolidine-4-r-carboxylate, methyl r-+-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate, methyl r-+-3-tert-butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylate, methyl r-n-boc-2,2-dimethyloxazolidine-4-carboxylate, r-+-3-boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, r-+-3-tert-butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylic acid methyl ester, r---3-boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, r-3-tert-butyl 4-methyl 2,2-dimethyloxazolidine-3,4-dicarboxylate | |
3-O-tert-butyl 4-O-methyl (4R)-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate |
Specifications
1.0820g/mL | |
Glass bottle | |
89.0°C (0.8 mmHg) | |
95715-86-9 | |
C12H21NO5 | |
3-tert-butyl 4-methyl 4r-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate, 3-tert-butyl 4-methyl r-+-2,2-dimethyloxazolidine-3,4-dicarboxylate, methyl 3-tert-butoxycarbonyl-2,2-dimethyloxazolidine-4-r-carboxylate, methyl r-+-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate, methyl r-+-3-tert-butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylate, methyl r-n-boc-2,2-dimethyloxazolidine-4-carboxylate, r-+-3-boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, r-+-3-tert-butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylic acid methyl ester, r---3-boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, r-3-tert-butyl 4-methyl 2,2-dimethyloxazolidine-3,4-dicarboxylate | |
CC1(N(C(CO1)C(=O)OC)C(=O)OC(C)(C)C)C | |
259.3 | |
259.3 | |
Methyl (R)-(+)-3-BOC-2,2-dimethyl-4-oxazolidinecarboxylate |
93°C | |
1.082 | |
1g | |
94% min. (GC) | |
MFCD00674041 | |
ZNBUXTFASGDVCL-MRVPVSSYSA-N | |
3-O-tert-butyl 4-O-methyl (4R)-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate | |
688220 | |
95% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning