Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl (R)-(-)-2-pyrrolidinone-5-carboxylate, 98%

$98.84 - $396.28

Chemical Identifiers

Chemical Name or Material Methyl (R)-(-)-2-pyrrolidinone-5-carboxylate
CAS 64700-65-8
Molecular Formula C6H9NO3
Formula Weight 143.14
MDL Number MFCD01318621
Synonym H-D-Pyr-OMe; Methyl D-pyroglutamate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3123603

  SDS
alfa aesar™
H3123603
1g Each for $98.84

AAH3123606

  SDS
alfa aesar™
H3123606
5g Each for $396.28
Description & Specifications

Specifications

Formula Weight 143.14
CAS 64700-65-8
Chemical Name or Material Methyl (R)-(-)-2-pyrrolidinone-5-carboxylate
Assay 98%
Boiling Point 83°C (0.25mmHg)
Synonym H-D-Pyr-OMe; Methyl D-pyroglutamate
Recommended Storage 2° to 8°C
Molecular Formula C6H9NO3
MDL Number MFCD01318621
Quantity 1g
Safety and Handling

2° to 8°C