Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl quinoxaline-6-carboxylate, 98%

$153.78 - $648.04

Chemical Identifiers

Chemical Name or Material Methyl quinoxaline-6-carboxylate
CAS 23088-23-5
Molecular Formula C10H8N2O2
Formula Weight 188.19
MDL Number MFCD00102618
Synonym Quinoxaline-6-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1668903

 
alfa aesar™
A1668903
1g Each for $153.78

AAA1668906

 
alfa aesar™
A1668906
5g Each for $648.04
Description & Specifications

Specifications

Formula Weight 188.19
Chemical Name or Material Methyl quinoxaline-6-carboxylate
Assay 98%
Molecular Formula C10H8N2O2
MDL Number MFCD00102618
CAS 23088-23-5
Synonym Quinoxaline-6-carboxylic acid methyl ester
Health Hazard 1 Warning
Melting Point 96° to 99°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.