Learn More
Methyl pyruvate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1396636
Description
Methyl Pyruvate acts as a potent secretagogue used in the study of stimulus-secretion coupling. Also used in the preparation of quinoxalines as dual phoosphodiesterase 2/10 (PDE2/10) inhibitors. It increases free calcium ion concentration in the cytosol of pancreatic cells. It is used in the preparation of dimethyl 2,3-dimethylenebutanedioate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl Pyruvate acts as a potent secretagogue used in the study of stimulus-secretion coupling. Also used in the preparation of quinoxalines as dual phoosphodiesterase 2/10 (PDE2/10) inhibitors. It increases free calcium ion concentration in the cytosol of pancreatic cells. It is used in the preparation of dimethyl 2,3-dimethylenebutanedioate.
Solubility
Soluble in chloroform, methanol, ether, alcohol. Slightly soluble in water.
Notes
Store away from oxidizing agent, heat.
Specifications
Methyl pyruvate | |
1.123 | |
39°C (102°F) | |
Sweet | |
1.404 | |
500 g | |
1361953 | |
methyl pyruvate, pyruvic acid, methyl ester, pyruvic acid methyl ester, methyl 2-oxopropionate, propanoic acid, 2-oxo-, methyl ester, methyl-pyruvate, methylglyoxylic acid methyl ester, unii-3kjm65g5xl, 3kjm65g5xl, 2-oxo-propionic acid methyl ester | |
CWKLZLBVOJRSOM-UHFFFAOYSA-N | |
methyl 2-oxopropanoate | |
11748 | |
102.09 | |
Liquid |
600-22-6 | |
135°C to 137°C | |
98% | |
C4H6O3 | |
MFCD00008754 | |
UN3272 | |
14,8021 | |
Soluble in chloroform,methanol,ether,alcohol. Slightly soluble in water. | |
CC(=O)C(=O)OC | |
102.089 | |
CHEBI:51850 | |
98% |
Chemical Identifiers
600-22-6 | |
102.089 | |
CWKLZLBVOJRSOM-UHFFFAOYSA-N | |
11748 | |
methyl 2-oxopropanoate |
C4H6O3 | |
MFCD00008754 | |
methyl pyruvate, pyruvic acid, methyl ester, pyruvic acid methyl ester, methyl 2-oxopropionate, propanoic acid, 2-oxo-, methyl ester, methyl-pyruvate, methylglyoxylic acid methyl ester, unii-3kjm65g5xl, 3kjm65g5xl, 2-oxo-propionic acid methyl ester | |
CHEBI:51850 | |
CC(=O)C(=O)OC |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P333+P313-P363-P370+P378q-P501c
H226-H317-H318-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 209-987-4
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only