missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl pyrrole-3-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H5434006

 View more versions of this product

Catalog No. AAH5434006



Description

Description

Applications
Methyl pyrrole-3-carboxylate is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Methyl pyrrole-3-carboxylate
5g
C6H7NO2
1h-pyrrole-3-carboxylic acid methyl ester, 1h-pyrrole-3-carboxylic acid, methyl ester, 3-methoxycarbonyl-1h-pyrrole, 4,5-methoxy-carbonylpyrrole, acmc-1cmqn, methyl 3-pyrrolecarboxylate, methyl pyrrole 3-carboxylate, methyl pyrrole-3-carboxylate, methylpyrrole-3-carboxylate, pubchem22507
WLBNVSIQCFHAQB-UHFFFAOYSA-N
methyl 1H-pyrrole-3-carboxylate
12361759
97%
85°C to 89°C
2703-17-5
MFCD00967074
Slightly soluble in water.
COC(=O)C1=CNC=C1
125.127
125.13

Chemical Identifiers

2703-17-5
125.127
WLBNVSIQCFHAQB-UHFFFAOYSA-N
12361759
COC(=O)C1=CNC=C1
C6H7NO2
MFCD00967074
1h-pyrrole-3-carboxylic acid methyl ester, 1h-pyrrole-3-carboxylic acid, methyl ester, 3-methoxycarbonyl-1h-pyrrole, 4,5-methoxy-carbonylpyrrole, acmc-1cmqn, methyl 3-pyrrolecarboxylate, methyl pyrrole 3-carboxylate, methyl pyrrole-3-carboxylate, methylpyrrole-3-carboxylate, pubchem22507
methyl 1H-pyrrole-3-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents