Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl pyridazine-4-carboxylate, 96%

$183.82 - $557.50

Chemical Identifiers

Chemical Name or Material Methyl pyridazine-4-carboxylate
CAS 34231-77-1
Molecular Formula C6H6N2O2
Formula Weight 138.13
MDL Number MFCD09953488
Synonym Pyridazine-4-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2744906

  SDS
alfa aesar™
H2744906
5g Each for $557.50

AAH2744903

  SDS
alfa aesar™
H2744903
1g Each for $183.82
Description & Specifications

Specifications

Formula Weight 138.13
Quantity 5g
Chemical Name or Material Methyl pyridazine-4-carboxylate
Melting Point 64° to 67°C
Molecular Formula C6H6N2O2
CAS 34231-77-1
Assay 96%
MDL Number MFCD09953488
Synonym Pyridazine-4-carboxylic acid methyl ester

Applications
It is used to produce 4-Pyridazincarbohydroxamsaeure by heating and requires a reagent hydroxylammoniumchloride and Na2CO3. Meanwhile, it needs solvent methanol.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.