Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl pyrazine-2-carboxylate, 98+%

$112.05 - $1,522.35

Chemical Identifiers

Chemical Name or Material Methyl pyrazine-2-carboxylate
CAS 6164-79-0
Molecular Formula C6H6N2O2
Formula Weight 138.13
MDL Number MFCD00014611
Beilstein 118345
Merck Index 14,7959
Synonym Methyl pyrazinoate; Pyrazine-2-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1202806

 
alfa aesar™
A1202806
5g Each for $112.05

AAA1202814

 
alfa aesar™
A1202814
25g Each for $410.99

AAA1202822

 
alfa aesar™
A1202822
100g Each for $1,522.35
Description & Specifications

Specifications

Formula Weight 138.13
Chemical Name or Material Methyl pyrazine-2-carboxylate
TSCA TSCA
Assay ≥98%
Molecular Formula C6H6N2O2
MDL Number MFCD00014611
CAS 6164-79-0
Beilstein 118345
Merck Index 14,7959
Synonym Methyl pyrazinoate; Pyrazine-2-carboxylic acid methyl ester
Health Hazard 1 Warning
Melting Point 56° to 60°C
Quantity 5g
Flash Point >100°C (212°F)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.