missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl Purple Solution pH 4.8(Purple) - pH 5.4(Gray) - pH 5.8(Green), TCI America™

$67.48 - $337.41

Chemical Identifiers

MDL Number MFCD00070843
Synonym Fleisher′s Indicator Solution
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M0112500ML View Documents Tci America
M0112500ML
500mL Each for $337.41
 
M011225ML View Documents Tci America
M011225ML
25mL Each for $67.48
 
Specifications

Chemical Identifiers

MFCD00070843
Fleisher′s Indicator Solution

Specifications

Green
MFCD00070843
Liquid at 20°C
500mL
Fleisher′s Indicator Solution
Methyl Purple Solution pH 4.8(Purple) - pH 5.4(Gray) - pH 5.8(Green)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator