Please login to your online account to display your discounted pricing

Methyl Purple Solution (Certified), Fisher Chemical


Quantity 500mL
Packaging Glass Bottle

Manufacturer: fisher scientific company  FLSI9-500

 View more versions of this product

Catalog No. SI9-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Quantity 500mL
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Color Dark Green
Packaging Glass Bottle
CAS 3486-30-4
CAS 67-63-0
CAS 7732-18-5
Dye Content Pass test
MDL Number MFCD00070843
ChemAlert Storage Symbol Red
Visual Transition Interval Pass test
Chemical Name or Material Methyl Purple Solution
CAS 845-10-3

Specifications

Chemical Name or Material Methyl Purple Solution
CAS 845-10-3
CAS 3486-30-4
CAS 67-63-0
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00070843
Accessories
 

SDS & Safety Documents