Please login to your online account to display your discounted pricing

Methyl Purple Solution (Certified), Fisher Chemical

$179.69 - $179.69

Chemical Identifiers

Chemical Name or Material Methyl Purple Solution
CAS 845-10-3
CAS 3486-30-4
CAS 67-63-0
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00070843
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SI9-500

  SDS
fisher scientific company
FLSI9-500
Glass Bottle 500mL Each for $179.69
Case of 6 Each for $1,078.14
Description & Specifications

Specifications

Purity Grade Certified Grade
Quantity 500mL
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Color Dark Green
Packaging Glass Bottle
CAS 67-63-0
CAS 3486-30-4
ChemAlert Storage Symbol Red
MDL Number MFCD00070843
Visual Transition Interval Pass test
Dye Content Pass test
CAS 7732-18-5
CAS 845-10-3
Chemical Name or Material Methyl Purple Solution

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Safety and Handling