Please login to your online account to display your discounted pricing

Methyl propyl disulfide, 80%, Tech., ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl propyl disulfide
Name Note tech., 80%
CAS 2179-60-4
Purity Grade Technical Grade
Assay ≥75%
Assay Percent Range 80%
Molecular Formula C4H10S2
Linear Formula CH3CHCHSSCH3
Formula Weight 122.25
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC337690050

  SDS
thermo fisher scientific
337690050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥75%
Purity Grade Technical Grade
Physical Form Liquid
Formula Weight 122.25
Quantity 5g
Color Clear slightly yellow
EINECS Number 218-551-2
TSCA TSCA
Density 0.98g/mL
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Methyl propyl disulfide
Infrared Spectrum Authentic
Refractive Index 1.5050 to 1.5090 (20°C, 589nm)
Name Note tech., 80%
Assay Percent Range 80%
Molecular Formula C4H10S2
Linear Formula CH3CHCHSSCH3
CAS 2179-60-4
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.