missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl propionate, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A14944AE

 View more versions of this product

Catalog No. AAA14944AE



Description

Description

Applications
It is commonly used in organic synthesis. It undergoes vapor-phase aldol condensation with formaldehyde to form methyl methacrylate.

Solubility
Soluble in water (5g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

Methyl Propionate
100mL
99%
MFCD00009306
1737628
methyl propionate, propanoic acid, methyl ester, methyl propylate, methylpropionate, propionic acid, methyl ester, propionate de methyle, propanoic acid methyl ester, fema number 2742, propionic acid methyl ester, methyl propionate natural
CCC(=O)OC
88.106
88.11
99%
0.915
554-12-1
C4H8O2
UN1248
14,6112
RJUFJBKOKNCXHH-UHFFFAOYSA-N
methyl propanoate
11124
Liquid

Chemical Identifiers

554-12-1
88.106
RJUFJBKOKNCXHH-UHFFFAOYSA-N
11124
CCC(=O)OC
C4H8O2
MFCD00009306
methyl propionate, propanoic acid, methyl ester, methyl propylate, methylpropionate, propionic acid, methyl ester, propionate de methyle, propanoic acid methyl ester, fema number 2742, propionic acid methyl ester, methyl propionate natural
methyl propanoate
Safety and Handling

Safety and Handling

GHS H Statement
H225-H332

GHS P Statement
P210-P261-P280-P240-P241-P303+P361+P353-P304+P340-P312-P403+P235-P501a

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL PROPIONATE

RTECSNumber : UF5970000

TSCA : TSCA

Documents